Sodium Hydroxide, Pellets (SQ), Qualigens
Qualigens

Sodium Hydroxide, Pellets (SQ), Qualigens

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 kg
50 kg
Packaging:
View all
Plastic Bottles
Catalog number Q27815
HSN 28151190
Price (INR)
659.00
Each
Quantity:
500 g
Packaging:
Plastic Bottles
Request bulk or custom format
Price (INR)
659.00
Each
Specifications
Cleanup SymbolCaustic
ColorWhite
Boiling Point1390°C
CAS1310-73-2
ChemAlert Storage SymbolWhite
View more

Detailed specifications PDF attached below