Sodium Lauryl Sulphate (SQ), Qualigens
Qualigens

Sodium Lauryl Sulphate (SQ), Qualigens

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 kg
Packaging:
Plastic Drum
Catalog number Q27825
HSN 34023900
Price (INR)
1,936.00
Each
Quantity:
500 g
Packaging:
Plastic Drum
Request bulk or custom format
Price (INR)
1,936.00
Each
Specifications
ColorWhite / pale yellow crystals /crystalline powder
CAS151-21-3
ChemAlert Storage SymbolRed
Health Hazard 2DANGER!
Health Hazard 3Emergency Overview
Flammable solid. Causes eye, skin, and respiratory tract irritation. May cause allergic respiratory reaction. Avoid contact with skin and eyes. Do not breathe dust. Use explosion-proof equipment. Use only non-sparking tools. Wash off immediately with soap and plenty of water removing all contaminated clothes and shoes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Remove from exposure, lie down. Remove from exposure, lie down. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0
View more

High purity and solubility ensures maximum interaction with water insoluble drugs. Also helps ensure an optimum level of drug release in media.

Detailed specifications PDF attached below