Product Image
Qualigens

Tetrabutylammonium Hydroxide 40%, for Ion Chromatography, Qualigens

Have Questions?
Catalog NumberQuantity
Q31263100 mL
Catalog number Q31263
HSN 29239000
Price (INR)
8,684.00
Each
Quantity:
100 mL
Request bulk or custom format
Price (INR)
8,684.00
Each
Specifications
Boiling Point97°C
Chemical Name or MaterialTetrabutylammonium hydroxide solution 40% in water
CAS2052-49-5
ColorColorless to very faint yellow liquid
Physical FormLiquid
View more
  • Used as a Regeneration Solution in cation Non suppressed applications on Ion chromatography
  • Used as Reagent for ion-pair extractions
  • Detailed specifications PDF attached below