Qubit™ RNA IQ Assay Kits
Invitrogen™

Qubit™ RNA IQ Assay Kits

Obtain ready-to-use detection of RNA degradation information with the Qubit RNA integrity and quality (IQ) Assay Kit and Standards, which feature two unique dyes for binding large and/or highly structured RNA and small, degraded RNA. Together, these dyes enable detection of RNA quality and integrity.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
Q33222275 assays
Q3322175 assays
Catalog number Q33222
Price (INR)
44,311.00
Online offer
Ends: 31-Dec-2026
63,301.00
Save 18,990.00 (30%)
Each
Quantity:
275 assays
Price (INR)
44,311.00
Online offer
Ends: 31-Dec-2026
63,301.00
Save 18,990.00 (30%)
Each

Quickly and easily determine RNA degradation with the Qubit RNA IQ Assay Kit and Qubit RNA IQ Standards. RNA integrity and quality (IQ) information is assessed using the assay kit's two unique dyes: one dye that binds to large and/or highly structured RNA and a second dye that selectively binds to small, degraded RNA. Together, the dyes enable you to quickly assess the quality and integrity of your RNA samples, with results presented as an RNA IQ number and as a calculated percent of large and small RNA in the sample.

Qubit RNA Integrity and Quality (IQ) Assay Kit provides a fast, simple method to check whether an RNA sample has degraded using a Qubit 4 or Qubit Flex fluorometer. The RNA IQ assay utilizes two unique dyes: one that binds to large, intact, and/or highly structured RNA (mRNA, tRNA, rRNA), and another that selectively binds to small and/or degraded RNA. Together they enable you to quickly assess the quality and integrity of the RNA sample.

To use, simply add your samples to the RNA IQ working solution, then measure the signal on a Qubit 4 or Qubit Flex fluorometer. Results are presented as an RNA IQ number (RNA IQ#) that indicates the RNA sample integrity and quality. Results are also posted as a calculated percent of large and small RNA in the sample. The RNA IQ# is a value from 1 to 10, similar to other RNA quality scores, where a small number indicates that the sample is comprised of mainly small RNA and a larger number indicates that the sample consists of mainly large RNA or RNA with tertiary structure.

Other features of the Qubit RNA IQ Assay Kit include:
• 5S and tRNA tertiary structure binds large RNA dye
• Accuracy even in the presence of salts, protein, solvents, and RNA stabilization reagents
• Signal stable for 1 hour

Solution-based chemistry in line with electrophoresis
The RNA IQ assay results correlate well with electrophoretic methods. The primary difference between the solution-based RNA IQ assay and traditional electrophoresis-based techniques is the amount of time it takes to ascertain whether an RNA sample has degraded. Gels are considered easy to perform, but still take time. Microfluidic-based methods not only take time to obtain results, but often have lengthy procedures to prepare the samples. Gel or microfluidic-based electrophoresis is still recommended for obtaining fragment size information or size distribution of the RNA sample.

Qubit RNA IQ Assay Standards
Qubit RNA IQ Assay RNA standards are designed for use with the Qubit RNA IQ Assay Kit. Sufficient amounts of the small and large and/or structured RNA standard are supplied for generating an IQ standard curve for up to 500 samples. The RNA IQ standards are designed to measure RNA degradation and are not interchangeable with the standards from other RNA quantitation assays, including the Qubit RNA HS, BR, or XR assay kits.

Notes
• Qubit RNA IQ assay kits are designed to work on Qubit 4 and Qubit Flex fluorometers
• The assay is not designed for use with Qubit, Qubit 2.0, or Qubit 3 fluorometers
• Use thin-wall, clear, 0.5 mL PCR tubes (Cat. No. Q32856) for the Qubit 4 Fluorometer and 8 x 200 μL tube strips (Cat. No. Q33252) for the Qubit Flex Fluorometer

For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayQubit RNA Integrity and Quality (IQ) Assay Kit
For Use With (Equipment)Qubit 4 Fluorometer, Qubit Flex Fluorometer
No. of Reactions275 Assays
Product LineQubit
Quantity275 assays
Shipping ConditionDry Ice
FormatKit
Sample TypeRNA
Unit SizeEach

Frequently asked questions (FAQs)

Can the Qubit RNA IQ Assay Kits (Cat. No. Q33221, Q33222) be run on a plate reader for high throughput screening of RNA integrity?

Qubit RNA IQ Assay Kits are not compatible with plate readers. The IQ value calculations are performed by the Qubit instrument, which involves proprietary algorithms that cannot be replicated on other devices. For accurate results, use the Qubit instrument as recommended.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I am using the Qubit RNA IQ Assay Kit (Cat. No. Q33221, Q33222). How do I convert the RNA IQ score to the RIN score?

There is no simple way to convert the RNA IQ score to RIN, because they measure different things. The application note, Qubit RNA IQ Assay: a fast and easy fluorometric RNA quality assessment, shows a detailed comparison and explanation of the two measurements. Figures 2 and 4 are most relevant.

Does the Qubit RNA IQ Assay detect free nucleotides/bases?

No. The presence of an excess amount of free nucleotides/bases may affect the assay results in the same manner as other contaminants, but the reagents in this kit do not give a fluorescence response to nucleotides or bases.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Do you offer a Qubit assay to detect purity of the RNA or DNA samples (level of protein contamination)?

At present, we do not have a fluorescence-based assay that tests for sample purity (protein contamination). We do offer various instruments that can obtain absorbance readings (absorbance at 260/280), such as our NanoDrop, BioMate and GENESYS spectrophotometers.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Does the Qubit RNA IQ Assay detect the purity of the RNA sample, similar to using the 260/280 absorbance method?

No. The Qubit RNA IQ Assay does not assay the purity of the RNA sample, only the extent of degradation of the RNA (small versus large strands).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (7)

Citations & References
Abstract
Anti-Inflammatory Effects of 5a,8a-Epidioxycholest-6-en-3ß-ol, a Steroidal Endoperoxide Isolated from Aplysia depilans, Based on Bioguided Fractionation and NMR Analysis.
Authors:Pereira RB, Pereira DM, Jiménez C, Rodríguez J, Nieto RM, Videira RA, Silva O, Andrade PB, Valentão P,
Journal:Mar Drugs
PubMed ID:31163615
'Sea hares of Aplysia genus are recognized as a source of a diverse range of metabolites. 5a,8a-Endoperoxides belong to a group of oxidized sterols commonly found in marine organisms and display several bioactivities, including antimicrobial, anti-tumor, and immunomodulatory properties. Herein we report the isolation of 5a,8a-epidioxycholest-6-en-3ß-ol (EnP(5,8)) from Aplysia depilans ... More
An Immune-Responsive Cytoskeletal-Plasma Membrane Feedback Loop in Plants.
Authors:Sassmann S, Rodrigues C, Milne SW, Nenninger A, Allwood E, Littlejohn GR, Talbot NJ, Soeller C, Davies B, Hussey PJ, Deeks MJ,
Journal:Curr Biol
PubMed ID:29937351
'Cell wall appositions (CWAs) are produced reactively by the plant immune system to arrest microbial invasion through the local inversion of plant cell growth. This process requires the controlled invagination of the plasma membrane (PM) in coordination with the export of barrier material to the volume between the plant PM ... More
Circular RNA circ-PRMT5 facilitates non-small cell lung cancer proliferation through upregulating EZH2 via sponging miR-377/382/498.
Authors:Wang Y, Li Y, He H, Wang F
Journal:Gene
PubMed ID:31479715
'Emerging evidence demonstrates that circular RNA (circRNA) is a novel class of non-coding RNA that plays a pivotal role in cancer. Recently, circ-PRMT5 was identified as an oncogene in bladder cancer. Nevertheless, its contribution to non-small cell lung cancer (NSCLC) is unknown. Herein, we aimed to clarify the biological role ... More
Microarray-Based Multivariate Analysis of the Effectiveness of Sublingual Immunotherapy for Cedar Pollinosis.
Authors:Gotoh M, Kaminuma O, Hiroi T, Okubo K,
Journal:Allergy Asthma Immunol Res
PubMed ID:30088375
Sublingual immunotherapy (SLIT) is an effective treatment for allergic diseases. However, the mechanism by which this therapy exhibits its efficacy has not been fully delineated. To elucidate the mechanisms of SLIT in the treatment of cedar pollinosis (CP), we performed a multivariate analysis of microarray data on mRNA expression in ... More
Porcine Circovirus Type 2 Induces Single Immunoglobulin Interleukin-1 Related Receptor (SIGIRR) Downregulation to Promote Interleukin-1ß Upregulation in Porcine Alveolar Macrophage.
Authors:Yang S, Liu B, Yin S, Shang Y, Zhang X, Khan MUZ, Liu X, Cai J
Journal:Viruses
PubMed ID:31684202
Multisystemic inflammation in pigs affected by porcine circovirus type 2 (PCV2) indicates the disordered expression of inflammatory cytokines. However, the PCV2-induced expression profile of inflammation cytokines and its regulating mechanism remain poorly understood. In this study, inflammatory cytokines and receptors in porcine alveolar macrophages (PAMs) after PCV2 infection were profiled ... More