Xylene, Sulphur Free (SQ), Qualigens
Qualigens

Xylene, Sulphur Free (SQ), Qualigens

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
25 L
200 L
Packaging:
View all
Glass Bottle
Catalog number Q35417
HSN 29024400
Price (INR)
2,776.00
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
2,776.00
Each
Specifications
Cleanup SymbolSolvent
Melting Point-34°C
CAS1330-20-7
ChemAlert Storage SymbolRed
Health Hazard 2WARNING!
View more

Detailed specifications PDF attached below