Silver Nitrate, N/10 (CVS), Qualigens
Qualigens

Silver Nitrate, N/10 (CVS), Qualigens

Have Questions?
Catalog NumberQuantityPackaging
Q371033 AmpulesPlastic Ampoules
Catalog number Q37103
HSN 28432100
Price (INR)
42,337.00
Each
Quantity:
3 Ampules
Packaging:
Plastic Ampoules
Request bulk or custom format
Price (INR)
42,337.00
Each
Specifications
Boiling Point444°C
Melting Point212°C
CAS7761-88-8
ChemAlert Storage SymbolYellow
DOT InformationDOT Class 5.1: Oxidizer
View more

This concentrated volumetric solution, which needs to be diluted, is easy to use and saves valuable shelf space.

Detailed specifications PDF attached below