Methyl Orange, Indicator, Solution, Qualigens
Qualigens

Methyl Orange, Indicator, Solution, Qualigens

Have Questions?
Catalog NumberQuantity
Q37833125 mL
Catalog number Q37833
HSN 38229090
Price (INR)
281.00
Each
Quantity:
125 mL
Request bulk or custom format
Price (INR)
281.00
Each
Specifications
Melting Point300°C
CAS547-58-0
ChemAlert Storage SymbolBlue
DOT InformationDOT Class 6.1, : Poison
Molecular FormulaC14H14N3O3S
View more

Detailed specifications PDF attached below