n-Butyl Alcohol, for HPLC, Qualigens
Qualigens

n-Butyl Alcohol, for HPLC, Qualigens

Have Questions?
Catalog NumberQuantityPackaging
Q431061 LGlass Bottle
Catalog number Q43106
HSN 29051300
Price (INR)
3,222.00
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
3,222.00
Each
Specifications
Cleanup SymbolSolvent
Melting Point-89°C
CAS71-36-3
ChemAlert Storage SymbolRed
Acidity0.002% Max (as CH3COOH)
View more

HPLC mobile phase for analytical and preparative separations

Detailed specifications PDF attached below