Water, for HPLC, Qualigens
Qualigens

Water, for HPLC, Qualigens

Have Questions?
Catalog NumberQuantityPackaging
Q439561 LPlastic Bottle
Catalog number Q43956
HSN 28530010
Price (INR)
729.00
Each
Quantity:
1 L
Packaging:
Plastic Bottle
Request bulk or custom format
Price (INR)
729.00
Each
Specifications
Absorbance0.005 Max [max. (1cm cell vs H2O) at 254nm]
pH5 to 8
CAS7732-18-5
ChemAlert Storage SymbolGray
CAS Min %0.99
View more

HPLC mobile phase for analytical and preparative separations

Detailed specifications PDF attached below