QuantiGene™ Plex Hybridization Plates
Invitrogen™
QuantiGene™ Plex Hybridization Plates
QuantiGene Plex Hybridization Plates are used for the overnight hybridization step of the QuantiGene Plex assay. This plate is aRead more
Have Questions?
Catalog NumberQuantity
QP054510 Plates
Catalog number QP0545
Price (INR)
14,574.00
Online offer
Ends: 31-Mar-2026
24,290.00
Save 9,716.00 (40%)
Each
Quantity:
10 Plates
Price (INR)
14,574.00
Online offer
Ends: 31-Mar-2026
24,290.00
Save 9,716.00 (40%)
Each
QuantiGene Plex Hybridization Plates are used for the overnight hybridization step of the QuantiGene Plex assay. This plate is a clear, polypropylene 96-well plate with round-bottom wells. Use with QuantiGene Plex Pressure Seals (Cat. No. QP0544) only.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypePCR Microplate
Quantity10 Plates
Shipping ConditionRoom Temperature
ColorClear
MaterialPolypropylene
No. of Wells96
Product LineQuantiGene Plex
Well ShapeRound Bottom (U-well)
Unit SizeEach