Human Promyelocytic Leukemia (HL-60) Total RNA for QuantiGene Assays
Invitrogen™
Human Promyelocytic Leukemia (HL-60) Total RNA for QuantiGene Assays
Human Promyelocyte Leukemia (HL-60) Total RNA is a total RNA sample extracted from HL-60 cells for use as a positiveRead more
Have Questions?
Catalog number QS0607
Price (INR)
15,424.00
Online offer
Ends: 30-Sep-2026
28,042.00
Save 12,618.00 (45%)
10 µg
Price (INR)
15,424.00
Online offer
Ends: 30-Sep-2026
28,042.00
Save 12,618.00 (45%)
10 µg
Human Promyelocyte Leukemia (HL-60) Total RNA is a total RNA sample extracted from HL-60 cells for use as a positive control in QuantiGene assays.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeHL-60 Cell
For Use With (Application)Nucleic Acid Quantitation
Product TypeTotal RNA
Quantity10 μg
Sample SourceTumor or Diseased Tissue, Cell Line
Shipping ConditionDry Ice
Sample TypeRNA
SpeciesHuman
Unit Size10 µg