Product Image
Invitrogen™

AP Enhancer Solution

AP Enhancer Solution, 10 mL is an aqueous buffered solution and an individual component of the ViewRNA™ ISH Tissue AssayRead more
Have Questions?
Catalog NumberQuantity
QVT050810 mL
Catalog number QVT0508
Price (INR)
10,547.00
Online offer
Ends: 31-Mar-2026
17,578.00
Save 7,031.00 (40%)
Each
Quantity:
10 mL
Price (INR)
10,547.00
Online offer
Ends: 31-Mar-2026
17,578.00
Save 7,031.00 (40%)
Each
AP Enhancer Solution, 10 mL is an aqueous buffered solution and an individual component of the ViewRNA™ ISH Tissue Assay Kits, 2-Plex, 24 assays. It is an enhancer solution for alkaline phosphatase.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeAP Enhancer Solution
Quantity10 mL
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C