Product Image
Invitrogen™
Probe Set Diluent QT
Probe Set Diluent QT, 12 mL is an aqueous solution containing formamide, detergent, and blocker. It is an individual componentRead more
Have Questions?
Catalog NumberQuantity
QVT051012 mL
Catalog number QVT0510
Price (INR)
17,103.00
Each
Quantity:
12 mL
Price (INR)
17,103.00
Each
Probe Set Diluent QT, 12 mL is an aqueous solution containing formamide, detergent, and blocker. It is an individual component of the ViewRNA™ ISH Tissue Assay Kits, 2-Plex, 24 assays.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeProbe Set Diluent
Quantity12 mL
FormatKit
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C