Product Image
Invitrogen™

Label Probe Diluent QF

Label Probe Diluent QF, 24 mL, is an aqueous solution containing detergent and an individual component of the ViewRNA™ ISHRead more
Have Questions?
Catalog NumberQuantity
QVT051324 mL
Catalog number QVT0513
Price (INR)
10,547.00
Online offer
Ends: 31-Mar-2026
17,578.00
Save 7,031.00 (40%)
Each
Quantity:
24 mL
Price (INR)
10,547.00
Online offer
Ends: 31-Mar-2026
17,578.00
Save 7,031.00 (40%)
Each
Label Probe Diluent QF, 24 mL, is an aqueous solution containing detergent and an individual component of the ViewRNA™ ISH Tissue Assay Kits, 2-Plex, 24 assays.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Recommended StorageStore at 2°C to 8°C
Quantity24 mL
Unit SizeEach