TheraPure™ Nucleotides
Thermo Scientific™

TheraPure™ Nucleotides

Thermo Scientific TheraPure in vitro transcription (IVT) enzymes are designed for advanced research and early development of mRNA therapeutics and vaccines.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeConcentration
R00046T1GTP Solution, Tris Solution200 mM
R044T1ATP Solution, Sodium Solution100 mM
R00044T1ATP Solution, Tris Solution200 mM
R045T1CTP Solution, Sodium Solution100 mM
R00045T1CTP Solution, Tris Solution200 mM
R046T1GTP Solution, Sodium Solution100 mM
R047T1UTP Solution, Sodium Solution100 mM
R00047T1UTP Solution, Tris Solution200 mM
R0491T1N1-Methylpseudo-UTP, Sodium Solution100 mM
Catalog number R00046T1
Price (INR)
5,271.00
Online offer
Ends: 31-Mar-2026
8,785.00
Save 3,514.00 (40%)
Each
Product Type:
GTP Solution, Tris Solution
Concentration:
200 mM
Price (INR)
5,271.00
Online offer
Ends: 31-Mar-2026
8,785.00
Save 3,514.00 (40%)
Each
Thermo Scientific TheraPure in vitro transcription (IVT) enzymes are designed for advanced research and early development of mRNA therapeutics and vaccines. These products offer full workflow solutions from template preparation to RNA synthesis and are recommended for process research and optimization, proof of concept, and preclinical studies.

The high purity of TheraPure IVT enzymes is ensured by precise quality testing that includes confirmation that that the enzymes are nuclease- and endotoxin-free. In addition, the manufacturing process of TheraPure products follows ISO9001 and ISO13485 standards.

Features

  • Convenient stand-alone enzymes for easy workflow optimization
  • Animal-origin-free (AOF) raw materials used in manufacturing
  • No risk of prion or animal virus contamination
  • Reproducible and consistent results with validated quality testing
  • Regulatory compliant—β-lactam-free manufacturing technology
  • Suitable for translational research
  • Facilitates transition to GMP during mRNA vaccine development process
  • Scalable and available as OEM product

The exceptional quality of TheraPure products is supported by:

  • Certificates of analysis
  • Certificates of origin
  • TSE/BSE statement
For Research Use Only.
Specifications
ColorColorless
For Use With (Application)In vitro Transcription
Product TypeGTP Solution, Tris Solution
Quantity0.2 mL
Shipping ConditionDry Ice
Concentration200 mM
Product LineTheraPure
Unit SizeEach
Contents & Storage
Store at -20°C.