Product Image
Thermo Scientific™

Remel™ Nitrate Medium

Thermo Scientific™ Remel Nitrate Medium is for the determination of nitrate reduction by Cryptococcus spp.
Have Questions?
Catalog NumberQuantity
R06154220 x 2 mL
Catalog number R061542
Price (INR)
-
Quantity:
20 x 2 mL

Thermo Scientific™ Remel Nitrate Medium is for the determination of nitrate reduction by Cryptococcus spp.

Requires:

Nitrate Reagent A, Part No. R21239 and Nitrate Reagent B, Part No. R21242.

Specifications
DescriptionNitrate Medium
Format15mm x 103mm Tube
Quantity20 x 2 mL
Product TypeMicrobiology Media
Unit SizeEach