Owl™ S3S Aluminum-Backed Sequencer System Replacement Parts
Actual product may vary

Owl™ S3S Aluminum-Backed Sequencer System Replacement Parts

Achieve optimal manual sequencing with Thermo Scientific™ Owl™ S3S Aluminum-Backed Sequencer System Replacement Parts.
Have Questions?
Catalog NumberDescription
R10383Buffer drain bottle
Catalog number R10383
HSN 90279090
Price (INR)
-
Description:
Buffer drain bottle

Achieve optimal manual sequencing with Thermo Scientific™ Owl™ S3S Aluminum-Backed Sequencer System Replacement Parts, for use with the Owl™ S3S Aluminum-Backed Sequencer System.

Choose from these replacement parts for optimal manual sequencing.
Specifications
DescriptionBuffer drain bottle
For Use With (Equipment)Owl™ S3S Aluminum-Backed Sequencer System
Product TypeDrain Bottle
Unit SizeEach