Plate Count Agar
Actual product may vary
Plate Count Agar
Plate Count Agar
Thermo Scientific™
Plate Count Agar
Enumerate viable organisms in milk, water, food and dairy products using this non-selective medium.
Have Questions?
Catalog NumberQuantityFormatMedia TypeManufacturing Origin
R11258910/Pk.Bottle - 100mLPlate Count AgarUnited States
Catalog number R112589
Price (INR)
-
Quantity:
10/Pk.
Format:
Bottle - 100mL
Media Type:
Plate Count Agar
Manufacturing Origin:
United States
Enumerate viable organisms in milk, dairy products, water, food and other specimens of sanitary importance using non-selective Thermo Scientific™ Plate Count Agar.
This standard medium meets the formulation of APHA, AOAC and PHLS.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
FormatBottle - 100mL
Media TypePlate Count Agar
Quantity10/Pk.
Manufacturing OriginUnited States
Product TypeAgar
Unit SizeEach