Product Image
Thermo Scientific™

Remel™ Colorimeter Standard Set, Vitek™

Thermo Scientific™ Remel Colorimeter Standard Kit contains latex equivalence turbidity standards for use with Vitek colorimeter.
Have Questions?
Catalog NumberFor Use With (Equipment)
R20343Vitek
Catalog number R20343
Price (INR)
-
For Use With (Equipment):
Vitek

Thermo Scientific™ Remel Colorimeter Standard Kit contains latex equivalence turbidity standards for use with Vitek colorimeter.

Contains one each of 0.5, 1.0, 2.0, and 3.0 standards.
Specifications
DescriptionColorimeter Standard Set
Includesone each of 0.5, 1.0, 2.0, and 3.0 standards
For Use With (Equipment)Vitek
Unit SizeEach