Remel™ Nitrate Swab
Remel™ Nitrate Swab
Remel™ Nitrate Swab
Remel™ Nitrate Swab
Thermo Scientific™
Remel™ Nitrate Swab
Thermo Scientific™ Remel Nitrate Swab is used for detection of nitrate utilization by yeast.
Have Questions?
Catalog NumberPackaging
R203556 Swabs/Pk.
Catalog number R20355
Price (INR)
-
Request A Quote
Packaging:
6 Swabs/Pk.
Thermo Scientific™ Remel Nitrate Swab is used for detection of nitrate utilization by yeast.
Requires:
Requires addition of Nitrate Reagent A, Part No. R21239 and Nitrate Reagent B, Part No. R21242.
Specifications
DescriptionNitrate Swab
Packaging6 Swabs/Pk.
Required AccessoriesRequires addition of Nitrate Reagent A, Part No. R21239 and Nitrate Reagent B, Part No. R21242.
Unit SizeEach