Remel™ BactiCard™ E. coli
Thermo Scientific™
Remel™ BactiCard™ E. coli
Thermo Scientific™ Remel BactiCard E. coli is used for presumptive identification of Escherichia coli using enzyme technology.
Have Questions?
Catalog NumberQuantity
R2110825 Tests/Kit
Catalog number R21108
Price (INR)
-
Quantity:
25 Tests/Kit
Thermo Scientific™ Remel BactiCard E. coli is used for presumptive identification of Escherichia coli using enzyme technology.
  • CPT Code: 87147
  • CLIA Complexity: Non-waived
  • Two separate test circles for MUG and Indole
  • Results after 15 minutes of incubation
  • Positive MUG test displays fluorescence under longwave ultraviolet light
  • Positive Indole test evidenced by a color change to redv
  • If indole and MUG are positive, then E. coli is present
Includes:
Test cards, rehydrating fluid, MUG reagent, and applicator sticks.
For in vitro diagnostic use.
Specifications
DescriptionBactiCard™ E. coli
Packaging Type1 Kit
Quantity25 Tests/Kit
Test Time15 min.
TypeBactiCard
Detectable AnalytesE. coli
Unit SizeEach
Contents & Storage
2°C to 8°C