Remel™ BactiCard™ Strep
Thermo Scientific™

Remel™ BactiCard™ Strep

Thermo Scientific™ Remel BactiCard Strep is used for presumptive identification of streptococci and related genera using enzyme technology.
Have Questions?
Catalog NumberQuantity
R2111225 Tests/Kit
Catalog number R21112
Price (INR)
-
Quantity:
25 Tests/Kit

Thermo Scientific™ Remel BactiCard Strep is used for presumptive identification of streptococci and related genera using enzyme technology.

Specifications
Control SetsPositive, Negative
DescriptionBactiCard™ Strep
For Use With (Application)Enzyme Immunoassay Detection
Packaging Type1 Kit
Quantity25 Tests/Kit
Target Organism ClassStaphylococcus aureus
TypeBacticard for Streptococcus Testing
CE MarkerYes
Detectable AnalytesClumping Factor and Protein A
Unit SizeEach