Remel™ Anaerobic Nitrate Reagent B
Thermo Scientific™
Remel™ Anaerobic Nitrate Reagent B
Thermo Scientific™ Remel Anaerobic Nitrate Reagent B is for determination of nitrate reduction in anaerobic bacteria.
Have Questions?
Catalog NumberQuantity
R21211-MBD25 mL
Catalog number R21211-MBD
Price (INR)
-
Quantity:
25 mL
Thermo Scientific™ Remel Anaerobic Nitrate Reagent B is for determination of nitrate reduction in anaerobic bacteria.
Ordering Alerts:
Used in conjunction with Anaerobic Nitrate Reagent A, Part No. R21201
Specifications
DescriptionAnaerobic Nitrate Reagent B
FormatBottle
Quantity25 mL
Detectable AnalytesNitrite, Free Nitrogen Gas
Unit SizeEach