Product Image
Thermo Scientific™
Remel™ Voges-Proskauer B Test
Determine the ability of gram-negative bacilli to produce acetylmethylcarbinol (acetoin) from glucose fermentation using Voges-Proskauer B Test.
Have Questions?
Catalog NumberQuantity
R2128125 mL
Catalog number R21281
Price (INR)
-
Quantity:
25 mL
Determine the ability of gram-negative bacilli to produce acetylmethylcarbinol (acetoin) from glucose fermentation using Thermo Scientific™ Remel™ 40% Potassium Hydroxide Reagent for Voges-Proskauer B Test. In 1898, Voges and Proskauer reported production of acetylmethylcarbinol from glucose fermentation by certain bacteria when grown in an appropriate medium such as MR-VP Broth1,2. Later Barret made the VP test more sensitive by using 40% Potassium Hydroxide (VP-B) to the medium for the detection of acetylmethylcarbinol production3.
The Voges-Proskauer test is based on the detection of acetylmethylcarbinol from glucose metabolism4.Ease of Differentiation: VP positive organisms produce a pink color development while VP negative organisms produce no color.Rapid Detection: Within 15 minutes.Glucose is metabolized to pyruvic acid from which the neutral products acetoin and 2,3-butanediol are formed. In the presence of oxygen and alkali, acetoin and 2,3-butanediol are oxidized to diacetyl, the reactant for the pink color produced in Voges-Proskauer test. VP-B is an oxidizing agent which hastens oxidation of acetoin to diacetyl.Used in conjunction with VP-A, Part No.R21200. For use with MRVP Broth.Not all products are available for sale in all territories. Please inquire.Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
  1. Voges O. and Proskauer B., 1898, Zeit. Hyg. 28:20-32.
  2. Clark W. M. and H.A. Lubs, 1915, J. Infect. Dis., 17:160-173.
  3. Barritt, M.M. 1936. J. Pathol. Bacteriol. 42:441-454.
  4. MacFaddin J. F., 2000, Biochemical Tests for identification of Medical Bacteria, 3rd Lippincott Williams & Wilkins, Philadelphia, PA.
Specifications
DescriptionVoges-Proskauer B
Packaging Type25 mL/Bottle
Quantity25 mL
Detectable AnalytesAcetylmethylcarbinol
Unit SizeEach
Contents & Storage
Store at Room Temperature Until Used