Product Image
Thermo Scientific™
Remel™ BactiDrop™ Desoxycholate
Differentiate between bile soluble Streptococcus pneumoniae and other insoluble alpha-hemolytic streptococci using BactiDrop™ Desoxycholate (10%).
Have Questions?
Catalog NumberQuantity
R2150850 x 0.75 mL
Catalog number R21508
Price (INR)
-
Quantity:
50 x 0.75 mL
Differentiate between bile soluble Streptococcus pneumoniae and other insoluble alpha-hemolytic streptococci using Thermo Scientific™ Remel™ BactiDrop™ Desoxycholate (10%). In 1900, Neufeld used a bile salt sodium desoxycholate to detect autolysis, that differentiated bile-soluble S. pneumoniae from bile-insoluble alpha-hemolytic streptococci1. Evans employed 10% bile to lyse broth cultures of pneumococci2. Hawn and Beebe evaluated the effectiveness of applying bile salts directly to a blood agar plate culture of pneumococci3.
Use Bactidrop Desoxychocolate to differentiate between bile soluble Streptococcus pneumoniae and other insoluble alpha-hemolytic streptococci.Ready-to-use: Liquid reagentEasy to interpret:Spot Test Positive Test - Colony disintegration within 30 minutesNegative Test - Colony remains intact within 30 minutesTube Test Positive Test - Clearing of Test suspension within 3 hours; Control suspension remains turbidNegative Test - Test and Control suspensions remain turbid within 3 hoursThe bile solubility test is based on the observation that when 10% desoxycholate is applied S. pneumoniae cells lyse, while other streptococci and gram-positive cocci do not. Bile salts cause a cell membrane derangment and also lower surface tension at the medium-membrane interface4,5.S. pneumoniae possesses an autocatalytic enzyme that lyses the organism’s own cell wall during cell division. This autolytic intracellular enzyme causes the organism to undergo rapid autolysis when cultivated on artificial medium6. Goebel and Avery stated that the action of bile salts is independent of the autolytic enzyme7. Dubos and Hirsch found that bile salts inactivate the naturally occurring autolytic enzyme either by the alteration or the removal of a normal autolytic inhibitor from the pneumococcal cells8.This reagent contains 10% w/v desoxycholic acid sodium, ethanol, sodium Merthiolate and demineralized water.Not all products are available for sale in all territories. Please inquire.Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
General References:
  1. Neufeld, F. 1900. Z. Hyg. Infektionskr. 34:454-464.
  2. Evans, A.C. 1936. J. Bacteriol. 31:423-437.
  3. Hawn, C.V.Z. and E. Beebe. 1965. J. Bacteriol. 90:549.
  4. Downie, A.W., L.Stent, and S.M. White. 1931. Br. J. Exp. Pathol. 12:1-9.
  5. Oginsky, E.L. and W.W. Umbreit. 1959. An Introduction to Bacterial Physiology. 2nd ed. W.H. Freeman and Co., San Francisco, CA.
  6. Cowan, S.T. 1974. Cowan & Steel's Manual for the Identification of Medical Bacteria. 2nd ed. Cambridge University Press, Cambridge, MA.
  7. Goebel, W.F. and O.T. Avery. 1929, J. Exp. Med. 49:267-286.
  8. Dubos, R.J. and J.G. Hirsch. 1965. Bacterial and Mycotic Infections of Man. 4th ed. Lippincott Co., Philadelphia, PA.
Specifications
DescriptionBactiDrop Desoxycholate
Packaging Type50/Pk.
Quantity50 x 0.75 mL
CE MarkerYes
Unit SizeEach
Contents & Storage
20°C to 25°C