Product Image
Thermo Scientific™
Remel™ Fecal Transport Two-Vial Sets: Cary-Blair w/Indicator/Empty Vial
Remel Fecal Transport Two-Vial Sets: Cary-Blair w/Indicator/Empty Vial offer preservation and recovery of enteric bacterial pathogens.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
R2161615 mL
R2194930 mL
Catalog number R21616
Price (INR)
-
Volume (Metric):
15 mL
Remel Fecal Transport Two-Vial Sets: Cary-Blair w/Indicator/Empty Vial offer preservation and recovery of enteric bacterial pathogens.
Caps are color coded to match industry standard colors and have an attached spork.
  • Broad range with long shelf life
  • Industry standard color-coded caps and labels
  • Easy to use spork cap for safe collection
  • Convenient packaging, including bulk kits
  • Multilingual patient collection instructions
  • Empty Vial is used for test procedures not compatible with fecal preservatives
  • Cary-Blair with Indicator contains 0.16% agar and is for preservation and recovery of enteric bacterial pathogens
Specifications
DescriptionCary-Blair/Empty Vial; Two-vial Sets
Packaging2-Vial Set
Volume (Metric)15 mL
Product TypeFecal Transport Set
Unit SizeEach