SAF Fixative Single Vial
Actual product may vary
Thermo Scientific™
SAF Fixative Single Vial
Collect, preserve, concentrate and permanently stain intestinal parasites with a quality fecal transport which comes with industry standard color-coding and sporked caps.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R21921120 Vials/Pk.
R2173012 Vials/Pk.
Catalog number R21921
Price (INR)
-
Quantity:
120 Vials/Pk.
Collect, preserve, concentrate and permanently stain intestinal parasites with Thermo Scientfic™ SAF Fixative Single Vial, part of a complete collection of quality fecal transports which come with industry standard color-coding and easy-to-use sporked caps.
Add convenience and safety to your fecal collection procedures with a broad line of transport products which have a long shelf life and are designed especially for enteric diagnostic testing. Look to us for convenient kit packaging options, including bulk kits. Multilingual patient collection instructions are included. Choose from a wide range of collection and preservation products for fecal specimens, which is important to the successful growth, isolation and detection of microorganisms.
  • Cary Blair with Indicator
  • Formalin 10%
  • Formalin 5%
  • PVA Modified Fixative
  • SAF Fixative
  • Zinc PVA Fixative
  • Empty Vial
Not all products are available for sale in all territories. Please inquire. Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
DescriptionSAF Fixative Single Vial
Fill Volume15 mL
PackagingSingle vial. Packaged in partitioned boxes.
Quantity120 Vials/Pk.
Product TypeSAF Fixative
Unit SizeEach