ProSpecT™ Adenovirus Microplate Assay
This product is no longer available to order. If you would like to discuss your options, or ask for a technical representative to call you, please contact Technical Support.
ProSpecT™ Adenovirus Microplate Assay
ProSpecT™ Adenovirus Microplate Assay
ProSpecT™ Adenovirus Microplate Assay
Thermo Scientific™
ProSpecT™ Adenovirus Microplate Assay
Rapidly detect Adenovirus specific antigen from human fecal samples.
Have Questions?
Catalog NumberQuantity
R24009696 Tests/Kit
Catalog number R240096
Price (INR)
-
Request A Quote
Quantity:
96 Tests/Kit
Rapidly detect Adenovirus specific antigen in fresh or preserved human fecal specimens using Thermo Scientific™ ProSpecT™ Adenovirus Microplate Assay.
Manage your resources, increase productivity and improve workflow efficiency with this direct, qualitative enzyme immmunoassay for the detection of Adenovirus. It combines excellent sensitivity/specificity, a common procedure with no indeterminate range, plus flexible specimen transport options.
  • Excellent clinical performance
  • Cary Blair transport option which saves a dilution step
  • Results within 100 minutes
  • Room temperature incubation
  • True colorless negatives
ProSpecT Adenovirus is part of the most comprehensive line of microplate assays available for enteric infectious disease testing. The ProSpecT family of tests have a common format, common procedures and common reagents, maximizing operational efficiency, productivity and throughput. ProSpecT Adenovirus provides same-day detection that is easy-to-use, flexible and provides the power to do more with less.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
DescriptionProSpecT Adenovirus Microplate Assay
IncludesTransfer pipettes, microplate strip holder and cover, microplate (8 wells/strip), enzyme conjugate, positive control, negative control, sample diluent, wash buffer, color substrate, stop solution and procedure card
Quantity96 Tests/Kit
Unit SizeEach
Contents & Storage
2°C to 8°C