ProSpecT™ Giardia Controls
Thermo Scientific™
ProSpecT™ Giardia Controls
Rapidly detect Giardia specific antigen from human fecal samples.
Have Questions?
Catalog NumberQuantity
R242804 Vials/Pk.
Catalog number R24280
Price (INR)
-
Quantity:
4 Vials/Pk.
Rapidly detect Giardia specific antigen in fresh or preserved human fecal specimens using Thermo Scientific™ ProSpecT™ Giardia Microplate Assays with ProSpecT™ Giardia Controls. Package contains two positive and two negative control vials for use with Giardia assays.
Manage your resources, increase productivity and improve workflow efficiency with a direct, qualitative enzyme immmunoassay for the detection of Giardia. It combines excellent sensitivity/specificity, a common procedure with no indeterminate range, plus flexible specimen transport options.
  • Excellent clinical performance
  • Cary Blair transport option which saves a dilution step
  • Results within 100 minutes
  • Room temperature incubation
  • True colorless negatives
ProSpecT Giardia is part of the most comprehensive line of microplate assays available for enteric infectious disease testing. The ProSpecT family of tests have a common format, common procedures and common reagents, maximizing operational efficiency, productivity and throughput. ProSpecT Giardia provides same-day antigen detection that is easy-to-use, flexible and provides the power to do more with less.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
DescriptionProSpecT Giardia Controls
For Use With (Application)Enzyme Immunoassay Detection
IncludesTwo positive and two negative control vials (0.75mL)
Quantity4 Vials/Pk.
Sample TypeFecal
TypeGiardia Antigen Testing
Detectable AnalytesGiardia
Unit SizeEach
Contents & Storage
2°C to 8°C