Product Image
Thermo Scientific™
Remel™ Xpect™ Giardia/Cryptosporidium Control Kit
Use in qualitative procedures as external control material for the Xpect Cryptosporidium, Xpect Giardia, or Xpect Giardia/Cryptosporidium test kits.
Have Questions?
Catalog NumberQuantity
R24505303 Vials/Kit
Catalog number R2450530
Price (INR)
-
Quantity:
3 Vials/Kit
Remel Xpect Giardia/Cryptosporidium Control Kit is used in qualitative procedures as external control material for the Xpect Cryptosporidium, Xpect Giardia, or Xpect Giardia/Cryptosporidium test kits.
Includes:
1 Giardia positive vial, 1 Cryptosporidium positive vial and 1 negative vial
Specifications
DescriptionXpect™ Giardia/Cryptosporidium Control Kit
Quantity3 Vials/Kit
CE MarkerYes
Detectable AnalytesGiardia and Cryptosporidium Antigen
Unit SizeEach
Contents & Storage
2°C to 8°C