ProSpecT™ Cryptosporidium Microplate Assay
Thermo Scientific™
ProSpecT™ Cryptosporidium Microplate Assay
Rapidly detect Crytosporidium specific antigen from human fecal samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R245409696 Wells/Kit
R245402424 Wells/Kit
Catalog number R2454096
Price (INR)
-
Quantity:
96 Wells/Kit
Rapidly detect Crytosporidium specific antigens (CSA) in fresh or preserved fecal specimens using Thermo Scientific™ ProSpecT™ Cryptosporidium Microplate Assay.
Manage your resources, increase productivity and improve workflow efficiency with this direct, qualitative enzyme immmunoassay for the detection of Crytosporidium. It combines excellent sensitivity/specificity, a common procedure with no indeterminate range, plus flexible specimen transport options.
  • Excellent clinical performance
  • Cary Blair transport option which saves a dilution step
  • Results within 100 minutes
  • Room temperature incubation
  • True colorless negatives
ProSpecT Cryptosporidium is part of the most comprehensive line of microplate assays available for enteric infectious disease testing. The ProSpecT family of tests have a common format, common procedures, and common reagents, maximizing operational efficiency, productivity and throughput.
ProSpecT Cryptosporidium provides same-day antigen detection that is easy-to-use, flexible and provides the power to do more with less.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
DescriptionProSpecT Cryptosporidium Microplate Assay
IncludesTransfer pipettes, microplate strip holder and cover, microplate (8 wells/strip), enzyme conjugate, positive control, negative control, sample diluent, wash buffer, color substrate, stop solution and procedure card
Quantity96 Wells/Kit
Detectable AnalytesCryptosporidium Antigen
Unit SizeEach
Contents & Storage
2°C to 8°C