Remel™ Color Slide™ II Mononucleosis Test
Thermo Scientific™
Remel™ Color Slide™ II Mononucleosis Test
Rapid slide agglutination test using horse erythrocytes for detection of heterophile antibodies of infectious mononucleosis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionQuantity
R2468936Color Slide™ II Mononucleosis Test24 Tests
R2471088Color Slide™ II Mononucleosis Test150 Tests
R2468944Color Slide™ II Mononucleosis Test50 Tests
Catalog number R2468936
Price (INR)
-
Description:
Color Slide™ II Mononucleosis Test
Quantity:
24 Tests
Thermo Scientific™ Remel Color Slide and II Mononucleosis Test is a rapid slide agglutination test using horse erythrocytes for detection of heterophile antibodies of infectious mononucleosis.
  • Dual color indicator
Specifications
DescriptionColor Slide™ II Mononucleosis Test
Quantity24 Tests
Sample TypePlasma, Serum
TypeReagent
Detectable AnalytesMononucleosis Antibodies
Unit SizeEach
Contents & Storage
2°C to 8°C