ProSpecT™ Campylobacter Microplate Assay
ProSpecT™ Campylobacter Microplate Assay
ProSpecT™ Campylobacter Microplate Assay
Thermo Scientific™
ProSpecT™ Campylobacter Microplate Assay
Rapidly detect Campylobacter specific antigen from human fecal samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R247609696 Tests/Kit
R247604848 Tests/Kit
Catalog number R2476096
Price (INR)
-
Quantity:
96 Tests/Kit
Rapidly detect Campylobacter specific antigen in fresh or preserved human fecal specimens using Thermo Scientific™ ProSpecT™ Campylobacter Microplate Assay.
Manage your resources, increase productivity and improve workflow efficiency with this direct, qualitative enzyme immmunoassay for the detection of Campylobacter. It combines excellent sensitivity/specificity, a common procedure with no indeterminate range, plus flexible specimen transport options.
  • Excellent clinical performance
  • Cary Blair transport option which saves a dilution step
  • Results within 100 minutes
  • Room temperature incubation
  • True colorless negatives
ProSpecT Campylobacter is part of the most comprehensive line of microplate assays available for enteric infectious disease testing. The ProSpecT family of tests have a common format, common procedures and common reagents, maximizing operational efficiency, productivity and throughput. ProSpecT Campylobacter provides same-day antigen detection that is easy-to-use, flexible and provides the power to do more with less.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Quantity96 Tests/Kit
TypeCampylobacter Microplate Assay
CE MarkerYes
Detectable AnalytesCampylobacter Antigen
Unit SizeEach
Contents & Storage
2°C to 8°C