Product Image
Thermo Scientific™
Remel™ Agglutinating Sera, Salmonella H
Used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
R30161701Salmonella H (k)
R30161601Salmonella H (i)
R30160401Salmonella H (c )
R30160201Salmonella H (a)
R30161401Salmonella H (gq)
R30160901Salmonella H (enz15)
R30160301Salmonella H (b)
R30160501Salmonella H (d)
R30161301Salmonella H (gp)
R30162901Salmonella H (z29)
R30163101Salmonella H (z38)
R30163001Salmonella H (z36)
R30162201Salmonella H (r )
R30161501Salmonella H (gst)
R30161901Salmonella H (lv)
R30162601Salmonella H (z6)
R30162801Salmonella H (z27)
R30162101Salmonella H (mt)
R30160101Rapid Diagnostic 3 (d, E, G, k)
R30161801Salmonella H (L) Polyvalent
R30160601Salmonella H (E) Polyvalent
R30162501Salmonella H (z4, z23)
R30162401Salmonella H (z)
R30163201Polyvalent, Phase 2
R30959801Rapid Diagnostic 1 (b, d, E, r)
R30162701Salmonella H (z10)
R30959901Rapid Diagnostic 2 (b, E, k, L)
R30162001Salmonella H (lw)
Catalog number R30161701
Price (INR)
-
Description:
Salmonella H (k)
Remel Agglutinating Sera, Salmonella H is used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
  • Select sera also for use as positive controls for stained Salmonella suspensions
Specifications
Control SetsPositive
DescriptionSalmonella H (k)
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella H
TypeAgglutinating Sera
Detectable AnalytesSalmonella H Antigen
Unit SizeEach