Search
Search
| Catalog Number | Description |
|---|---|
| R30161901 | Salmonella H (lv) |
| R30161601 | Salmonella H (i) |
| R30160401 | Salmonella H (c ) |
| R30160201 | Salmonella H (a) |
| R30161401 | Salmonella H (gq) |
| R30160901 | Salmonella H (enz15) |
| R30160301 | Salmonella H (b) |
| R30160501 | Salmonella H (d) |
| R30161301 | Salmonella H (gp) |
| R30162901 | Salmonella H (z29) |
| R30163101 | Salmonella H (z38) |
| R30163001 | Salmonella H (z36) |
| R30162201 | Salmonella H (r ) |
| R30161701 | Salmonella H (k) |
| R30161501 | Salmonella H (gst) |
| R30162601 | Salmonella H (z6) |
| R30162801 | Salmonella H (z27) |
| R30162101 | Salmonella H (mt) |
| R30160101 | Rapid Diagnostic 3 (d, E, G, k) |
| R30161801 | Salmonella H (L) Polyvalent |
| R30160601 | Salmonella H (E) Polyvalent |
| R30162501 | Salmonella H (z4, z23) |
| R30162401 | Salmonella H (z) |
| R30163201 | Polyvalent, Phase 2 |
| R30959801 | Rapid Diagnostic 1 (b, d, E, r) |
| R30162701 | Salmonella H (z10) |
| R30959901 | Rapid Diagnostic 2 (b, E, k, L) |
| R30162001 | Salmonella H (lw) |