Product Image
Thermo Scientific™
Remel™ Agglutinating Sera, Vibrio cholerae
Used for serological identification of Vibrio cholerae by slide and/or tube agglutination test.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionDetectable AnalytesQuantity
R30165101Vibrio cholerae InabaV. cholerae2 mL
R30165201Vibrio cholerae OgawaV. cholerae2 mL
Catalog number R30165101
Price (INR)
-
Description:
Vibrio cholerae Inaba
Detectable Analytes:
V. cholerae
Quantity:
2 mL
Remel Agglutinating Sera, Vibrio cholerae are used for serological identification of Vibrio cholerae by slide and/or tube agglutination test.
Specifications
DescriptionVibrio cholerae Inaba
FormatVial
Packaging Type2 mL/Vial
Quantity2 mL
CE MarkerYes
Detectable AnalytesV. cholerae
Unit SizeEach
Contents & Storage
2°C to 8°C