Product Image
Thermo Scientific™
Remel™ Agglutinating Sera, Proteus O
Remel Agglutinating Sera, Proteus O are used as positive controls for stained Proteus suspensions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionQuantityDetectable Analytes
R30165901Proteus OXK2 mLProteus Suspension
R30165701Agglutinating Sera, Proteus O, OX2 Positive Controls2 mLProteus Suspension
R30165801Proteus OX192 mLProteus Suspension
Catalog number R30165901
Price (INR)
-
Description:
Proteus OXK
Quantity:
2 mL
Detectable Analytes:
Proteus Suspension
Remel Agglutinating Sera, Proteus O are used as positive controls for stained Proteus suspensions.
Specifications
DescriptionProteus OXK
FormatVial
Packaging Type2 mL/Vial
Quantity2 mL
CE MarkerYes
Detectable AnalytesProteus Suspension
Unit SizeEach
Contents & Storage
2°C to 8°C