Remel™ Agglutinating Sera, Haemophilus influenzae
Product Image
Thermo Scientific™
Remel™ Agglutinating Sera, Haemophilus influenzae
Used for serological identification of H. influenzae by rapid slide agglutination test and CIE procedures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
R30166001Haemophilus influenzae Type A
R30166401Haemophilus influenzae Type E
R30166501Haemophilus influenzae Type F
R30166101Haemophilus influenzae Type B
R30166201Haemophilus influenzae Type C
R30166301Haemophilus influenzae Type D
Catalog number R30166001
Price (INR)
-
Request A Quote
Description:
Haemophilus influenzae Type A
Remel Agglutinating Sera are used for serological identification of H. influenzae by rapid slide agglutination test and CIE procedures.
Specifications
DescriptionHaemophilus influenzae Type A
TypeAgglutinating Sera
Detectable AnalytesH. influenzae
Unit SizeEach