Product Image
Thermo Scientific™

Neisseria meningitidis Agglutinating Sera

Meningococcus Agglutinating Sera are used in the qualitative serological identification of Neisseria meningitidis Groups A, B, C, D, X, Y, Z, W135, A-D and X-Z + W135.

Have Questions?
Catalog NumberQuantity
R301666012 mL
Catalog number R30166601
Price (INR)
-
Quantity:
2 mL

Perform serological identification of Neisseria meningitidis Groups A, B, C, D, X, Y, Z and W135 for epidemiological and diagnostic purposes using Thermo Scientific™ Remel™ Meningococcus Agglutinating Sera. Group specificity is based on the identity of capsular polysaccharides and strains possessing these materials are specifically agglutinated by the homologous antiserum. In cases of bacterial meningitis it is important in to obtain a presumptive identification of the infecting agent as early as possible, so that adequate antimicrobial therapy can be started.

Serological tests are based on the fact that antibodies in serum, produced in response to exposure to bacterial antigens, will agglutinate with bacteria carrying homologous antigens. Homologous cultures should be used for positive control organisms.

  • Ready to use - Prepared reagents
  • Easy to store - Store at 2-8°C
  • Polyvalent sera are for slide use only.
  • Easy to interpret -
    • Test suspension - Strong and clearly visible agglutination within one minute.
    • Control suspension - No visible agglutination.

Meningococcus Agglutinating Serum is produced in rabbits and preserved with 0.5% phenol. Each dropper bottle contains 2 mL liquid and is ready for use.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications
DescriptionNeisseria meningitidis Polyvalent Groups A - D
MaterialPolyvalent
Organism GroupPolyvalent Groups A-D
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassNeisseria Meningitidis
TypeNeisseria Meningitidis Test Kit
CE MarkerYes
Detectable AnalytesN. meningitidis
Unit SizeEach