Meningococcus Agglutinating Sera
Actual product may vary
Thermo Scientific™
Meningococcus Agglutinating Sera
Serologically identify Neisseria meningitidis by rapid agglutination test.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberOrganism Group
R30167101Group X
R30166701Polyvalent Groups X-Z, W135
R30167201Group Y
R30167301Group Z
Catalog number R30167101
Price (INR)
-
Organism Group:
Group X
Serologically identify Neisseria meningitidis Groups A, C, D, X, Y, Z and W135 for epidemiological and diagnostic purposes with Thermo Scientific™ Remel™ Meningococcus Agglutinating Sera.
Use Meningococcus Agglutinating Sera for presumptive identification of Neisseria meningitidis grown on culture media.
  • Simple, easy-to-use tests
  • Identifies major groups of Neisseria meningitidis serotypes
  • Provides accurate, reliable and rapid results
  • Serological tests are based on the fact that antibodies in serum, produced in response to exposure to bacterial antigens, will agglutinate with bacteria carrying homologous antigens.
    Serological tests are intended for screening purposes and should augment, not replace, culture procedures.
    The monovalent meningococcus agglutinating sera are suitable for use in slide agglutination tests. An alternative method of typing, counter immunoelectroophoresis is also described in literature1,2,3,4. The polyvalent agglutinating sera are for slide use only.
    Not all products are available for sale in all territories. Please inquire.
    Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
    For In Vitro Diagnostic Use.  
    For In Vitro Diagnostic Use.  
    General References:
    1. Grados, O. and Ewing, W.H. (1970). Antigenic relationship between Escherichia coli and Neisseria meningitidis. J. Infect. Dis., 122, 100.
    2. Myerowitz, R.L., Gordon, R.E. et al. (1973). Polysaccharides of the genus Bacillus cross-reactive with the capsular polysaccharides of Diplococcus pneumoniae type III, Haemophilus influenzae type b, and Neisseria meningitidis group A. Infect. Immun., 8, 896.
    3. Robbins, J.B., Myerowitz, R.L. et al. (1972). Enteric bacteria cross reactive with Neisseria meningitidis groups A and C and Diplococcus pneumoniae types I and III. Infect. Immun., 6, 651.
    4. Tobin, B.M. and Jones, D.M. (1972). Immunoelectroosmophoresis in the diagnosis of meningococcal infections. J. Clin. Path., 15, 583.
    Specifications
    DescriptionMeningococcus Agglutinating Sera
    IncludesMeningococcus Agglutinating Sera 1 dropper bottle
    Organism GroupGroup X
    Quantity2 mL
    Sample TypeSerum
    TypeAgglutinating Sera
    Detectable AnalytesN. gonorrhoeae
    Unit SizeEach
    Contents & Storage
    2°C to 8°C