Product Image
Thermo Scientific™
PHA Purified
Phytohaemagglutinin (PHA) is a mitogenic agent which is used in studies involving cell permeability.
Have Questions?
Catalog NumberQuantity
R308528011 Vial
Catalog number R30852801
Price (INR)
-
Quantity:
1 Vial
Carry out studies involving cell permeability with Thermo Scientific™ Remel™ Phytohaemagglutinin (PHA). PHA is a mitogenic agent derived from extracts of Phaseolus vulgaris seeds and has twin properties of causing erythroagglutination and of stimulating progressive lymphocyte mitosis in cell culture. PHA has proved to be beneficial in the study of the immune response, lymphocyte kinetics, and bone marrow dynamics, and for these purposes it is desirable to use a substance which has more closely reproducible qualities and known potency.
Purified phytohaemagglutinin is a freeze-dried, highly refined protein fraction of selected Phaseolus spp. seed extract in which the specific mitogenic activity and the mitogenic/ haemagglutinating activity ratio have been increased by a factor of about 100/1 during purification. Both agglutinating and mitogenic activities of PHA are found to be associated with the protein fractions of crude extracts, which are sufficiently alike in their physico-chemical properties, thus it is difficult to separate them. Relatively crude extracts selected for their ability to yield good mitotic pictures have proved to be useful in routine lymphocyte culture for chromosome studies.
  • Ready to use - Prepared reagent
  • Easy to store - Store at 2° to 8°C until use
  • Can be used in routine lymphocyte culture for chromosome studies
Purified phytohaemagglutinin contains a freeze-dried, highly refined protein fraction of selected Phaseolus spp. seed extract. Each bottle contains 2mg lyophilised phytohaemagglutinin (purified), dried from a small volume of buffered saline without preservative. The reagent may be reconstituted to any desired volume in sterile saline (if buffered, preferably to a pH between 6.5 and 8) or distilled water, using a sterile disposable hypodermic syringe.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
DescriptionPHA Purified
FormatVial
Quantity1 Vial
Unit SizeEach
Contents & Storage
2°C to 8°C