Remel™ PYR Disc with Reagent
Actual product may vary
Thermo Scientific™
Remel™ PYR Disc with Reagent
Use for presumptive identification of enterococci, Lancefield group A streptococci, and E. coli based on PYRase production.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R3085430150 Test
R30854401100 Tests
R21117225 Tests/Kit
Catalog number R30854301
Price (INR)
-
Quantity:
50 Test
Thermo Scientific™ Remel PYR Disk w/Reagent is used for presumptive identification of enterococci, Lancefield group A streptococci, and E. coli based on PYRase production.
Specifications
Quantity50 Test
CE MarkerYes
Unit SizeEach
Contents & Storage
2°C to 8°C