Wellcogen™ Neisseria meningitidis A, C, Y, W135 Rapid Latex Agglutination Test
Thermo Scientific™
Wellcogen™ Neisseria meningitidis A, C, Y, W135 Rapid Latex Agglutination Test
Rapidly detect antigens to Neisseria meningitidis groups A, C, Y, W135 present in human body fluids or blood cultures by latex agglutination.
Have Questions?
Catalog NumberQuantity
R3085920330 Tests/Kit
Catalog number R30859203
Price (INR)
-
Quantity:
30 Tests/Kit
Wellcogen™ N. meningitidis ACY W135 is a rapid latex test for use in the qualitative detection of antigen from Neisseria meningitidis (meningococcus) groups A, C, Y and W135, present in body fluids as a consequence of infection or in blood cultures. NOTE: Tests performed directly on clinical specimens are intended for screening purposes and should augment, not replace, culture procedures. Results must be used in conjunction with other data; e.g. symptoms, results of other tests, clinical impressions etc.
Use this rapid latex test in the qualitative detection of antigen from Neisseria meningitidis (meningococcus) groups A, C, Y and W135 present in body fluids, as a consequence of infection, or in blood cultures.
The reagent consists of polystyrene latex particles which have been coated with antibodies specific to meningococcus groups A, C, Y and W135 antigens. These latex particles agglutinate in the presence of sufficient homologous antigen.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
DescriptionWellcogen Neisseria meningitidis A, C, Y, W135 Test Kit
MaterialPolystyrene
Organism GroupGroup A, C, Y, W135
Quantity30 Tests/Kit
TypeNeisseria Meningitidis Test Kit
CE MarkerYes
Unit SizeEach
Contents & Storage
2°C to 8°C