Stained Proteus Suspensions
Thermo Scientific™
Stained Proteus Suspensions
Rapidly detect and quantify reckettsial antibodies in sera with this suspension.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDetection Method
R30953601Proteus OX2
R30953701Proteus OX19
R30953801Proteus OXK
Catalog number R30953601
Price (INR)
-
Detection Method:
Proteus OX2
Rapidly detect and quantify rickettsial antibodies in sera with Thermo Scientific™ Remel™ Stained Proteus Suspensions. Stained Proteus Suspensions are standardized, smooth suspensions of killed bacteria which have been stained to facilitate reading of agglutination tests.
Use Stained Proteus Suspensions for the quantitative detection of rickettsial antibodies in sera for epidemiological and diagnostic purposes in the investigation of rickettsial infections (Weil-Felix test)1, 2.
  • Simple, easy to use tests
  • Provide accurate, reliable results in as little as one minute
The tests are based on the fact that somatic constituents of some strains of Proteus are shared with some species of Rickettsia. Sera from people with some rickettsial infections therefore will agglutinate suspensions of the Proteus strains.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Detection MethodProteus OX2
Quantity5 mL
Unit SizeEach
Contents & Storage
2 - 8 °C