Quant-iT™ RiboGreen ReadyPlate
Quant-iT™ RiboGreen ReadyPlate
Quant-iT™ RiboGreen ReadyPlate
Quant-iT™ RiboGreen ReadyPlate
Invitrogen™
Quant-iT™ RiboGreen ReadyPlate
The Quant-iT RiboGreen ReadyPlate is a preloaded 96-well plate that offers a rapid, simplified, and highly sensitive method for RNA quantification. The ReadyPlate is prefilled with buffer and RiboGreen reagent, which exhibits bright green fluorescence upon binding to RNA or DNA.
Have Questions?
Catalog NumberQuantity
R327051 x 96-Well ReadyPlate
Catalog number R32705
Price (INR)
20,084.00
キャンペーン価格
Ends: 30-Sep-2026
28,691.00
Save 8,607.00 (30%)
Each
Quantity:
1 x 96-Well ReadyPlate
Price (INR)
20,084.00
キャンペーン価格
Ends: 30-Sep-2026
28,691.00
Save 8,607.00 (30%)
Each
The Quant-iT RiboGreen ReadyPlate is a preloaded 96-well plate that offers a rapid, simplified, and highly sensitive method for RNA quantification. The ReadyPlate is prefilled with buffer and RiboGreen reagent, which exhibits bright green fluorescence upon binding to RNA or DNA.
The streamlined assay format, along with prediluted standards, enables users to get to their results faster. Here, the user needs to simply hydrate the wells, add the provided standards and user samples, and incubate for two minutes prior to fluorescent measurement. This format allows users to quantify 1–200 ng of RNA (0.01–2 ng/μL when using 100 μL samples).
The Quant-iT RiboGreen ReadyPlate is ideal for measuring yields of RNA preparations for microarray experiments, reverse transcription PCR (RT-PCR), differential display PCR, northern blot analysis, S1 nuclease assays, RNase protection assays, cDNA library preparations, and high-throughput automated workflows.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypePre-filled Microplate
Quantity1 x 96-Well ReadyPlate
Detection MethodFluorescence
Final Product TypeRNA (Labeled)
FormatKit
Labeling TargetRNA (General)
Label or DyeOther Labels or Dyes
Sample TypeRNA
Unit SizeEach