ReadyProbes™ In Situ Hybridization (ISH) Blocking Solution (5X)
Invitrogen™

ReadyProbes™ In Situ Hybridization (ISH) Blocking Solution (5X)

ReadyProbes In Situ Hybridization (ISH) Blocking Solution minimizes non-specific background staining in chromogenic in situ hybridization (CISH) or fluorescence inRead more
Have Questions?
Catalog NumberQuantity
R37620100 mL
Catalog number R37620
Price (INR)
-
Quantity:
100 mL
ReadyProbes In Situ Hybridization (ISH) Blocking Solution minimizes non-specific background staining in chromogenic in situ hybridization (CISH) or fluorescence in situ hybridization (FISH) procedures. It is provided as a 5X solution that is diluted to 1X with distilled or deionized water. After the labeled DNA or RNA probe hybridization and wash steps, the sample is incubated with 1X Ready Probes ISH Blocking Solution for 30–60 minutes. 1X ReadyProbes ISH Blocking Solution can also be used as diluent for antibodies, avidin, streptavidin, or biotin during incubation steps.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration5X
Product LineReadyProbes
Product TypeIn Situ Hybridization Blocking Solution
Quantity100 mL
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
Unit SizeEach
Contents & Storage
Provided as 100-mL solution at 5X concentration in amber HDPE bottle.

Store at 2–8°C.

When stored as directed, product can be used for 6 months after receipt.

Frequently asked questions (FAQs)

What is the volume for one drop from the ReadyProbes dropper bottles?

The volume for one drop from the ReadyProbes dropper bottles is approximately 41 µL.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can the ReadyProbes In Situ Hybridization (ISH) Blocking Solution (5X) be used with PrimeFlow and ViewRNA labeling?

We do not recommend using the ReadyProbes In Situ Hybridization (ISH) Blocking Solution (5X) with PrimeFlow and ViewRNA labeling as the PrimeFlow and ViewRNA kits utilize a specific blocking reagent optimized for these kits.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.