ReadyProbes™ Solvent-Resistant Permanent Marking Pen
Invitrogen™
ReadyProbes™ Solvent-Resistant Permanent Marking Pen
The ReadyProbes Solvent-Resistant Permanent Marking Pen enables solvent-resistant writing on frosted glass microscope slides. Using the pen on other typesRead more
Have Questions?
Catalog NumberQuantity
R376235 pens
Catalog number R37623
Price (INR)
9,315.00
線上優惠
Ends: 30-Sep-2026
12,420.00
Save 3,105.00 (25%)
Each
Quantity:
5 pens
Price (INR)
9,315.00
線上優惠
Ends: 30-Sep-2026
12,420.00
Save 3,105.00 (25%)
Each
The ReadyProbes Solvent-Resistant Permanent Marking Pen enables solvent-resistant writing on frosted glass microscope slides.  Using the pen on other types of hard surfaces could produce writing that is not solvent-resistent.  When used on frosted glass microscope slides, ReadyProbes Solvent-Resistant Permanent Marking Pen is resistant to most solvents, including alcohols and xylene.  Be sure to let the ink dry before exposing to solvent.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Includes5 Pens
Quantity5 pens
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
Product LineReadyProbes
TypePermanent Marking Pen
Unit SizeEach
Contents & Storage
A set of 5 pens.
Store at room temperature.
When stored as directed, product can be used for 6 months after receipt.
Frequently asked questions (FAQs)
When would I use the ReadyProbes Solvent-Resistant Permanent Marking Pen versus a standard hydrophobic barrier pen?
We recommend using the ReadyProbes Solvent-Resistant Permanent Marking Pen if you will be subjecting your samples to any solutions containing alcohol or xylene.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.