Product Image
Thermo Scientific™
Lactophenol Aniline Blue
Microscopically examine fungi in wet mount preparations using Lactophenol Aniline Blue.
Have Questions?
Catalog NumberQuantity
R4002825 mL/Bottle
Catalog number R40028
Price (INR)
-
Quantity:
25 mL/Bottle
Microscopically examine fungi in wet mount preparations using Thermo Scientific™ Remel™ Lactophenol Aniline Blue. Linder recommended lactophenol as a mounting fluid for examination and preservation of fungi1. He reported that lactophenol kills organisms without causing cytoplasmic plasmolysis. Lactophenol Aniline Blue is recommended by Koneman, Roberts, and Wright for direct mounting and staining of fungi.2
Lactophenol Aniline Blue is a stain recommended for use in wet mount preparations for direct mounting and staining of fungi.
  • Ready to use: Liquid reagent
  • Easy identification: Yeast cells, mycelia and fruiting structures stain blue and are visible under a microscope
This reagent contains phenol which precipitates cytoplasmic proteins and inactivates enzyme systems within fungal cell. Glycerol prevents drying, allowing for examination of the stain 18-24 hours after preparation. Lactic acid is a clearing agent. Aniline blue is a dye which stains the hyaline fungal structures making them more distinct.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
  1. Linder, D.H. 1929. Science. 70:430.
  2. Koneman, E.W., G.D. Roberts, and S.F Wright. 1978. Practical Laboratory Microbiology. 2nd Williams & Wilkins, Baltimore, MD.
Specifications
PackagingEach
Quantity25 mL/Bottle
Volume (Metric)25 mL/Bottle
Unit SizeEach