Product Image
Thermo Scientific™
D'Antoni's Iodine
Detect protozoan cysts and helminth eggs and larvae in wet mount preparations using D'Antoni's Iodine Stain.
Have Questions?
Catalog number R40034
Price (INR)
-
Detect protozoan cysts and helminth eggs and larvae in wet mount preparations using Thermo Scientific™ Remel™ D'Antoni's Iodine stain. This is a nonspecific, contrast dye, added to fresh, unpreserved fecal specimens to aid in detection of helminth eggs and larvae and in differentiating protozoan cysts from host white blood cells1,2.
The diagnosis of intestinal parasitic infections is confirmed by the identification of parasites in fecal specimens. D’Antoni’s Iodine stains the nuclei of protozoan cysts, making them more visible. Helminth eggs and larvae are recognizable in wet mounts, though iodine staining occasionally obscures the internal morphologic details.
  • Ready-to-use: Liquid reagent supplied in 25mL dropper bottles
  • Easy identification: Stains the nuclei of protozoan cysts, making them more visible.
  • Easy-to-store: Store at room temperature (20-25°C)
D’Antoni’s Iodine preparation contains 15g iodine and 10g potassium iodide dissolved in demineralized water. This preparation is ready to use and does not require further dilution.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
  1. Garcia, L.S. 2010. Clinical Microbiology Procedures Handbook. 3rd ed. ASM Press, Washington D.C.
  2. Clinical and Laboratory Standards Institute (CLSI). 2005. Procedures for the Recovery and Identification of Parasites from the Intestinal Tract; Approved Guideline. 2nd ed. M28-A2. CLSI, Wayne, PA.
Specifications
DescriptionD'Antoni's Iodine
FormatBottle
Packaging Type25 mL/Bottle, Ea.
Quantity25 mL
Unit SizeEach