See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
Home
▸
Shop All Products
▸
Biochemical Reagents
▸
Staining Reagents
▸
Stains and Dyes
▸
Remel™ Gram Crystal Violet
Thermo Scientific™
Remel™ Gram Crystal Violet
Thermo Scientific™ Remel Gram Crystal Violet is a primary stain in Gram stain procedures.
Have Questions?
Change view
Catalog Number
Quantity
R40053
250 mL, 5/Pk.
R40052
250 mL
R40073
1 gal.
3 Options
Catalog number
R40053
Price (INR)
-
Request A Quote
Quantity:
250 mL, 5/Pk.
Thermo Scientific™ Remel Gram Crystal Violet is a primary stain in Gram stain procedures.
For qualitative procedures to differentiate gram-positive from gram-negative microorganisms
Selected Gram stains are CE marked
Specifications
Certifications/Compliance
CE Marked
Quantity
250 mL, 5/Pk.
Product Type
Crystal Violet
Volume (Metric)
5 x 250 mL